PFA tongs
Lieferant: SAVILLEX
700-850
SAVI700-850EA 489 EUR
SAVI700-850
PFA tongs
Zangen
Adjustable PFA tongs are moulded from the highest purity PFA resin for ultratrace metals use.
Service temperature range is –200 to +260 °C (–328 to +500 °F).
Ideal for transferring ICP glassware during cleaning. Tongs are 28 cm (11") long with three adjustable width positions.