Tork Foam Soap
Lieferant: Essity
470022
SCAA470022EA 125 EUR
SCAA470022
Tork Foam Soap
Hautpflege Seifen und Hautreiniger
Tork foam soap, with its high capacity, provides portion control and helps prevent overuse.
  • Perfumed fragrance, premium quality
  • Colour: Pink
  • Compatible with S34 dispensers