Tork W1/W2/W3 Light Duty Cleaning Cloths, Combi-Roll
Lieferant: Essity
500137
SCAA500137EA 92.7 EUR
SCAA500137
Tork W1/W2/W3 Light Duty Cleaning Cloths, Combi-Roll
Wischtücher Reinigungstücher, wiederverwendbar Tücher für allgemeine Anwendungen
Multipurpose cleaning cloths for light-duty cleaning tasks. Use these durable cloths wet or dry to clean dust and dirt, and can rinse and reuse them as needed.
  • These disposable cleaning cloths are compatible with most chemical solvents, so no need to change the cleaning products
  • This combi-roll has 600 sheets to avoid runouts and save refill time
Tork light-duty cleaning cloths are an economical, multipurpose wiper. Compatible with most chemical solvents, these durable cloths can be used wet or dry for various light-duty cleaning tasks. They’re soft and flexible, making it easy to clean in hard-to-reach places, and more durable than a paper wiper, so can rinse and reuse them. The combi-roll has a high capacity to save time on refills.
Standards: FSC MIX Credit, SA-COC-008266.