Tork® Extra Hand Sanitising Alcohol Gels
Lieferant: Essity
424205 511205 590205
SCAA424205EA 174 EUR
SCAA424205 SCAA511205 SCAA590205
Tork® Extra Hand Sanitising Alcohol Gels
Hautpflege Seifen und Hautreiniger
Tork extra hand sanitising alcohol gel effectively kills microorganisms to help prevent the spread of infections. The most effective gel in the Tork collection, with efficacy against bacteria, yeast and all viruses. Ideal for everyday hand hygiene. Suitable for use in healthcare. The light, refreshing gel is non-sticky on the hands and contains moisturising ingredients. Gentle to sensitive skin in an extended use-test under dermatological control.
  • Effective
  • Help prevent infections
  • Factory-sealed bottle
  • Quality: Premium
  • Colour: Transparent
Carefully crafted with high-performing ingredients. Reduce the risk of harmful micro-organisms, such as bacteria or viruses entering the body to cause illness. A factory-sealed bottle with a new pump for every refill helps reduce the risk of contamination.
Helps secure good hygiene: Factory-sealed bottle with a new pump for every refill helps reduce risk of contamination and protect the formulations all the way to the user
Reduce waste: Bottle is collapsible, leading to 70% less waste volume
Use biocides safely. Always read the label and product information before use
Dermatologically tested. Excellent skin tolerance with frequent use Effective against a variety of problem organisms - conforms with EN 1500, EN 12791 and EN 14476 (fully virucidal - including all enveloped viruses, such as: HIV, corona, influenza, rotavirus and norovirus)
Reduce time for cleaning staff: Certified effortless refilling in less than 10 sec