Tork® Washstation Dispenser
Lieferant: Essity
651420
SCAA651420EA 363 EUR
SCAA651420
Tork® Washstation Dispenser
Wischtuch-Spender
Durable, water-resistant design holds up well in wet, harsh environments.
  • High-capacity design requires fewer reflls
  • Hygienic hands-free dispensing
  • Easy to wipe or spray down without removing from wall
  • Easy mounting with clear backplate