Vacuum/gas manifolds with metering valves, KIMBLE®
Lieferant: DWK Life Sciences
SP216050-0005 SP216050-0000
SCERSP216050-0005EA 2720 EUR
SCERSP216050-0005 SCERSP216050-0000
Vacuum/gas manifolds with metering valves, KIMBLE®
Verteiler
This all-glass vacuum/gas manifold is for connecting inert gas and vacuum sources to sidearm stopcocks.
  • Fine control of vacuum and inert gases provided by metering groove cut into plug of each stopcock
  • Distance between stopcocks is 65 mm for the 3-position size and 100 mm for the 5-position size
Standards: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements