Centrifuge tubes, KIMAX®, KIMBLE®
Lieferant: DWK Life Sciences
SP45243-100 SP45243-200
SCERSP45243-100EA 1020 EUR
SCERSP45243-100 SCERSP45243-200
Centrifuge tubes, KIMAX®, KIMBLE®
Röhrchen Zentrifugenröhrchen
KIMAX® tube are designed for field use in testing petroleum.
  • Calibrated to contain
  • Scale and legend are durable white ceramic enamel
  • Short cone oil centrifuge tubes 152,4 mm