Culture tubes, disposable, KIMBLE®
Lieferant: DWK Life Sciences
SP60GM166
SCERSP60GM166EA 297 EUR
SCERSP60GM166
Culture tubes, disposable, KIMBLE®
Röhrchen Kulturröhrchen
Manufactured from 90 expansion soda-lime glass, KIMAX®.
These premium quality tubes feature sturdy, uniform bottoms and consistent lengths.
Standards: Glass conforming USP Type III requirements