Stopcock, three-way
Lieferant: DWK Life Sciences
SP845100-0004
SCERSP845100-0004EA 740 EUR
SCERSP845100-0004
Stopcock, three-way
Hähne
Made of 33 expansion, low extractable borosilicate glass.
  • Very good chemical and temperature resistance
  • 1:10 taper
  • Hollow glass plug
Three-way, double-oblique bore, precision ground stopcock.Lower vacuum chamber is not in the inline of flow and must be outgassed in a separate operation.
Standards: Confirms to USP Type I and ASTM E438, Type I, Class A requirements.