Microfiltration assembly, with solvent pick-up adapter, ULTRA-WARE®, KIMBLE®
Lieferant: DWK Life Sciences
SP953820-1047
SCERSP953820-1047EA 2030 EUR
SCERSP953820-1047
Microfiltration assembly, with solvent pick-up adapter, ULTRA-WARE®, KIMBLE®
Filter Membranfiltergeräte
This microfiltration system manufactured from 33 expansion, low extractable borosilicate glass eliminates pouring HPLC solvents into funnels. The solvent pickup adapter draws solvent directly from a reagent bottle through a 47 mm filter and into a filter flask.
  • Support base with 40 to 60 micron porosity glass frit
  • Integral vacuum connection above the drip tip to prevent contamination of the vacuum line with filtrate droplets
After filtration is complete, the HI-VAC valve on the solvent pickup adapter can be turned off for vacuum degassing.
Existing ULTRA-WARE® and Millipore filtration assemblies are easily converted to in-line filtration/degassing by replacing existing funnels with solvent pickup adapters. They clamp to support bases in the same way that funnels do.
Standards: Conforms to USP Type I and ASTM E438, Type I, Class A requirements
Lieferumfang: System supplied with a solvent pickup adapter, a coarse porosity fritted glass filter base, an inner joint flask and an aluminum clamp.