ORP electrode solutions
Lieferant: SI Analytics
285138184 285138357 285138373 285138381 285138168
662-6171EA 64 EUR
662-6171 SCOI285138357 SCOI285138373 SCOI285138381 SCOI285138168
ORP electrode solutions
Standards Redox-/ORP-Standards
ORP electrode solutions are used as cleaning solutions for combination electrodes and reference electrodes.