Flow through cooler CK 310
Lieferant: SI Analytics
285414320
SCOI285414320EA 4790 EUR
SCOI285414320
Flow through cooler CK 310
Umwälzthermostate Geschlossene Umwälzsysteme Kälteumwälzthermostate
The flow-through cooler CK 310 is the perfect complement to bath thermostats of CT 72 series to extending their temperature range to below ambient temperature.
  • Small footprint
  • Low energy requirement: Economic solution preventing tap water usage for cooling
  • Internal cooling loop is made of stainless steel: Better chemical resistance than copper