Organglas, DURAN®
Lieferant: DWK Life Sciences
242042409
SCOT242042409EA 300 EUR
SCOT242042409
Organglas, DURAN®
Dosen Probengläser
DURAN®, borosilicate glass 3.3, clear. Without stopper.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Organ storage jars, cylindrical vessels with a wide neck, are used to store materials.