Accessories for SCAT Politainer
Lieferant: S.C.A.T.
107338
SCTV107338EA 64.9 EUR
SCTV107338
Accessories for SCAT Politainer
Sicherheitsbehälter
The SCAT Politainer - also known as folding canisters - is a combination of a flexible plastic bag and a stable UN-approved outer carton. Storage in the unfilled state is possible in a very small space and after filling, safe storage or transport is guaranteed.
  • Thread to adjust the thread from GL38 to GL45
  • Especially suitable for SCAT Politainer
SCAT Politainers are an ideal way of not having to dispense with safe storage even in limited spaces.