Disinfectant, terralin® Liquid
Lieferant: SCHÜLKE & MAYR GMBH
102001
SCUE102001EA 252 EUR
SCUE102001
Disinfectant, terralin® Liquid
Reinigungsmittel Desinfektionsmittel
terralin® can be used for rapid disinfection of medical devices [CE0297] in all hospital areas with a high infection risk, e.g. treatment units and adjoining surfaces, examination tables, operating tables and adjoining surfaces.
  • Alcohol based rapid disinfectant, containing no aldehydes
  • Ready to use
  • Rapid and effective in many applications
  • Dries rapidly, leaving no streaks