Disinfectant liquid, Mikrozid® AF liquid
Lieferant: SCHÜLKE & MAYR GMBH
109301
SCUE109301EA 169 EUR
SCUE109301
Disinfectant liquid, Mikrozid® AF liquid
Reinigungsmittel Desinfektionsmittel
Alcohol-based, rapid disinfection of non-invasive medical devices and surfaces.
  • Ready to use, alcohol based disinfectant, aldehyde-free
  • Very broad biocidal activity within a short time
  • Dries rapidly without leaving smears
  • Good wetting properties