Thermosept® NKZ
Lieferant: SCHÜLKE & MAYR GMBH
146503
SCUE146503EA 71.9 EUR
SCUE146503
Thermosept® NKZ
Reinigungsmittel Desinfektionsmittel
Neutraliser based on citric acid for the automated reprocessing of medical devices and laboratory accessories.
  • Free of phosphates and surfactants
  • Gentle on material
  • Prevents the building of deposits
  • Optimises the cleaning results after neutral cleaning
  • Efficient neutralisation after alkaline cleaning
Thermosept® NKZ is used for the reprocessing of surgical and microsurgical instruments, stainless steel containers, rigid endoscopes, anaesthetics material, operating theatre shoes and baby bottles or laboratory equipment after alkaline cleaning, to avoid the spread of alkaline residues. It is also suitable for acidic pre-cleaning of stainless steel instruments. thermosept® NKZ can be used in washer/disinfectors and multi-chamber washer/disinfectors.