Dispenser for hand disinfectants and lotions, plastic, sm2
Lieferant: SCHÜLKE & MAYR GMBH
668500
SCUE668500EA 97.1 EUR
SCUE668500
Dispenser for hand disinfectants and lotions, plastic, sm2
Seifen- und Flüssigseifenspender
Wall-mountable plastic dispenser for use with hand disinfectants and lotions (500 and 1000 ml bottles).
  • No contamination and evaporation thanks to the unique pump design and integrated bottle cap
  • Valve protected against contact and contamination
  • Almost complete emptying of the container thanks to the flexible suction hose
  • Adjustable dosage stroke: 1, 2 or 3 ml
  • Easy cleaning and, if necessary, disinfection thanks to easy dismantling and complete dismantling of the dispenser and pump
  • Easy mounting