gigasept® AF Forte
Lieferant: SCHÜLKE & MAYR GMBH
70003412 70003413
SCUE70003412EA 966 EUR
SCUE70003412 SCUE70003413
gigasept® AF Forte
Reinigungsmittel Desinfektionsmittel
gigasept® AF forte is an aldehyde-free preparation for cleaning and disinfection of rigid and flexible endoscopes as well as surgical instruments.
  • Aldehyde-free
  • Suitable for flexible and rigid endoscopes
  • Shortened action time in ultrasonic baths
  • Patented substance combination
Cleaning and disinfection of flexible and rigid endoscopes, surgical instruments, anaesthesia equipment and breathing masks. Green colour, liquid form, pH 9,8 to 10.