Open top, 2 or 3 compartment dispensers
Lieferant: S-CURVE
BD-03
SCURBD-03EA 387 EUR
SCURBD-03
Open top, 2 or 3 compartment dispensers
Spender für PSA
Two- and three-compartment dispensers are made from clear, clean, 6 mm thick acrylic.
Open top provides easy access to items. Ideal for gloves, glove liners, face masks, and other disposables. Edges are flame-treated for smoothness. Rubber pads on bottom corners prevent slipping.