Recombinant Human Glutathione Peroxidase 2
Lieferant: SELENOZYME
The human GPX2 selenoprotein is encoded by the GPX2 gene and has potent hydrogen peroxide reducing activity using glutathione.
We provide the human GPX2 enzyme with approx. 20% Sec contents, mixed with inactive forms containing Sec-to-Lys and Sec-to-Gln substitutions, at more than 95% purity in aliquots at amounts from 0,5 to 5 units.
- Genuine Sec-containing selenoproteins
- No animals used as protein sources (grown in bacteria)
- More than 90% Sec contents
- More than 95% purity
Lieferumfang: Protein stored in 25 mM Tris-HCl (pH 8.0) with 250 mM KCl, 2 mM 2-Mercaptoethanol and 10% glycerol. Concentration 1 mg protein/ml. Shipped in cold pack by courier service. Store at −20 °C upon delivery.
Verpackung: Aliquots
Bitte beachten Centrifuge the tube briefly before opening the lid.