Recombinant Human Thioredoxin Reductase 2
Lieferant: SELENOZYME
The human TrxR2 selenoprotein is encoded by the TXNRD2 gene and exerts a wide range of functions in mitochondrial redox control and antioxidant systems. We provide the main mitochondrial isoform of the human TrxR2 enzyme with more than 90% Sec contents and more than 95% purity in aliquots at amounts from 5 to 50 units.
- Genuine Sec-containing selenoproteins
- No animals used as protein sources (grown in bacteria)
- More than 90% Sec contents
- More than 95% purity
Lieferumfang: Protein stored in TE buffer with 50% glycerol. Concentration 1 mg protein/ml. Shipped in cold pack by courier service. Store at −20 °C upon delivery.
Verpackung: Aliquots
Bitte beachten Centrifuge the tube briefly before opening the lid.