Recombinant Human Thioredoxin Reductase 2
Lieferant: SELENOZYME
HTRXR2-5 HTRXR2-10 HTRXR2-20 HTRXR2-50
SELZHTRXR2-5EA 750 EUR
SELZHTRXR2-5 SELZHTRXR2-10 SELZHTRXR2-20 SELZHTRXR2-50
Recombinant Human Thioredoxin Reductase 2
Proteine und Peptide
The human TrxR2 selenoprotein is encoded by the TXNRD2 gene and exerts a wide range of functions in mitochondrial redox control and antioxidant systems. We provide the main mitochondrial isoform of the human TrxR2 enzyme with more than 90% Sec contents and more than 95% purity in aliquots at amounts from 5 to 50 units.
  • Genuine Sec-containing selenoproteins
  • No animals used as protein sources (grown in bacteria)
  • More than 90% Sec contents
  • More than 95% purity
Lieferumfang: Protein stored in TE buffer with 50% glycerol. Concentration 1 mg protein/ml. Shipped in cold pack by courier service. Store at −20 °C upon delivery.
Verpackung: Aliquots
Bitte beachten Centrifuge the tube briefly before opening the lid.