Recombinant Rat Thioredoxin Reductase 1
Lieferant: SELENOZYME
RTRXR1-5 RTRXR1-10 RTRXR1-20 RTRXR1-50
SELZRTRXR1-5EA 750 EUR
SELZRTRXR1-5 SELZRTRXR1-10 SELZRTRXR1-20 SELZRTRXR1-50
Recombinant Rat Thioredoxin Reductase 1
Proteine und Peptide
The rat TrxR1 selenoprotein is encoded by the rat Txnrd1 gene and exerts a wide range of functions in cytosolic redox control and antioxidant systems.
We provide the main cytosolic isoform of the rat TrxR1 enzyme with more than 90% Sec contents and more than 95% purity in aliquots at amounts from 5 to 50 units.
  • Genuine Sec-containing selenoproteins
  • No animals used as protein sources (grown in bacteria)
  • More than 90% Sec contents
  • More than 95% purity
Lieferumfang: Protein stored in TE buffer with 50% glycerol. Concentration 1 mg protein/ml. Shipped in cold pack by courier service. Store at −20 °C upon delivery.
Verpackung: Aliquots
Bitte beachten Centrifuge the tube briefly before opening the lid.