Dropping bottle, LDPE
Lieferant: SEMADENI
0021
SEMA0021EA 3.5 EUR
SEMA0021
Dropping bottle, LDPE
Flaschen Tropfflaschen
LDPE, translucent.
  • Good chemical resistance
  • Good temperature resistance
Dropping bottle with a long tip and cap holder at the tape.
Bestellen
TECHNISCHE DATEN
Environmentally Preferable