Tubing connectors, X-shaped, PP
Lieferant: SEMADENI
0553
SEMA0553EA 5.8 EUR
SEMA0553
Tubing connectors, X-shaped, PP
Fittings und Verbinder
Tubing connectors, made of PP. Dimensions are shown as both nominal and exact as elasticity of tubing will influence the choice of size. Use our tubing selector 229-0111 to determine "grip". This will then give equivalent crest dimension for the particular connector concerned.
  • Good mechanical strength and chemical resistance
  • Very good thermal resistance
  • Autoclavable
  • Suitable for foodstuff