Sleeves for conical ground joints
Lieferant: SEMADENI
0614
SEMA0614EA 5.9 EUR
SEMA0614
Sleeves for conical ground joints
Normschliff-Manschetten und Faltenbälge Schliffmanschetten
Made of PTFE, white.
  • Very good chemical resistance
  • Gas, liquid and vacuum-tight (to 0,1 mbar)
  • Temperature resistant from −200 to +280 °C
For fitting onto standard ground cones. Easily detachable and suitable for repeated use. Joint grease is not needed.