Drums, UN approved, with screw lids
Lieferant: SEMADENI
0719
SEMA0719EA 240 EUR
SEMA0719
Drums, UN approved, with screw lids
Fässer, Fässer, Trommeln
Drums, made of blue HDPE.
  • Very sturdy
  • Good chemical resistance
  • Good temperature resistance
  • UN-approved