Newbox, PP
Lieferant: SEMADENI
12672
SEMA12672EA 0 EUR
SEMA12672
Newbox, PP
Boxen Lagerboxen
The closed Euro-standard container of the Newbox range boasts a slim design. Thanks to its robustness and dimensional stability, the Newbox is a durable and economic product.
  • Temperature resistant up to 80 °C (short term up to 100 °C)
  • 100% compatible with RAKO stacking containers
  • With two and four handle openings
The containers are stackable and dimensionally matched with Euro pallets. Compared with the Logiline range, the Newbox range impresses with a somewhat slimmer and lighter design that is nevertheless robust and dimensionally stable.
Applications: With manual handling closed Newbox containers are used universally as transport containers or for the storage of the widest possible range of goods.