Containers, rectangular
Lieferant: SEMADENI
1298 1297 1296 1294
SEMA1298EA 60.3 EUR
SEMA1298 SEMA1297 SEMA1296 SEMA1294
Containers, rectangular
Probenbehälter Universalbehälter
HDPE or PP, white, stackable.
These are ideal containers with side handles and edge for gripping to handle solid samples. Containers over 225 mm height are not suitable for liquids.