Waste Bin, PP
Lieferant: SEMADENI
1331
SEMA1331EA 81.6 EUR
SEMA1331
Waste Bin, PP
Behälter Abfallbehälter
Bins, made of PP.
  • Tough and dimensionally stable
  • Smooth surface facilitates easy cleaning
  • Stackable conical containers
These versatile and universally applicable plastic containers are suitable for sorting and storage of solid waste.