Polyurethane tubings, transparent
Lieferant: SEMADENI
2591
SEMA2591EA 13.5 EUR
SEMA2591
Polyurethane tubings, transparent
Schläuche Transferschlauch
Made of PU.
  • High mechanical strength
  • Resistant to dilute acids and alkaline solutions
Suitable for pressure lines, clean-up and analysis.
Operating temperature range −50 to 80 °C.