Safety Container
Lieferant: SEMADENI
3651
SEMA3651EA 259 EUR
SEMA3651
Safety Container
Transportbehälter Flaschentragebehälter
Safety container made of PC, with hermetically sealed lid and stainless steel mounting bracket.
  • Robust design
  • Avoid direct contact with solvents, strong acids, and alkaline solutions
Ideal for the safe storage and transport of test tube racks and small vessels with bacteriological and hazardous substance.