Wash bottles, wide neck, with coloured cap
Lieferant: SEMADENI
4580 4575 4576 4581
SEMA4580EA 29.3 EUR
SEMA4580 SEMA4575 SEMA4576 SEMA4581
Wash bottles, wide neck, with coloured cap
Flaschen Spritzflaschen
LDPE, with colour coded caps
  • No dripping during storage and use
  • easy refilling through wide neck opening
Wide neck, natural/translucent with color printed labels and warning symbols, with pressure relief valve.