Storage cabinet
Lieferant: SEMADENI
5187
SEMA5187EA 43.3 EUR
SEMA5187
Storage cabinet
Möbel Lagerschränke
The storage cabinet system allows a quick, individual and efficient division of drawers, shelves, cupboards, etc.
  • The boxes can be put together in a number of variations according to a sophisticated module
  • The boxes have holders on the bottom, which fit into the grid area of ​​the self-adhesive drawer foil
  • This self-adhesive foil prevents the boxes from slipping around in the drawer
  • Heavy-duty boxes made of impact-resistant polystyrene (PS).