Racks for test tubes
Lieferant: SEMADENI
5291
SEMA5291EA 13.9 EUR
SEMA5291
Racks for test tubes
Gestelle Reagenzglasgestelle
Test tube racks made of PP usable on both sides with individual frames connected together with or without optional carrying handle. 60 compartments for microtubes from 0,2 to 0,5 ml, 80 compartments for reaction tubes of Ø 10 to 12 mm and cryotubes or microtubes 1,5 to 2 ml.
  • Stackable
  • Good chemical resistance
  • Autoclavable up to 121 °C
  • Openings are numbered