Test tube racks made of PP usable on both sides with individual frames connected together with or without optional carrying handle. 60 compartments for microtubes from 0,2 to 0,5 ml, 80 compartments for reaction tubes of Ø 10 to 12 mm and cryotubes or microtubes 1,5 to 2 ml.