Developer Tray, PVC
Lieferant: SEMADENI
658
SEMA658EA 0 EUR
SEMA658
Developer Tray, PVC
Schalen/Schüsseln/Töpfe Laborschalen und Laborwannen
Made from white PVC, with grooved bottom, very sturdy and low-priced tray, acid-proof, temperature-resistant up to +60 °C.