AssetTag
Lieferant: SenseAnywhere
0096647494982
SENA0096647494982EA 15 EUR
SENA0096647494982
AssetTag
Datenlogger
An AssetTag is a card with barcode for the holder and QR code for on the outside of an asset representing the monitored asset in SAClient.
  • Monitor temp/RH of the asset
  • Data loggers can be switched at any time
  • Complete temp/RH history of assets without any monitoring-gaps
  • No reconfiguration needed (alarms, reports etc.)
  • No paperwork or administration
  • Not prone to error or hassle
  • Complete audit trail
How it works: The back of the card has two-sided foam tape. The card can be snapped into two parts. Bottom part: attach to holder in the asset. Top part: on outside of the asset (fridge/freezer). Scan the QR code to see the state of the asset. See which device (serial number) is attached. Proceed to SAClient for coupling, decoupling or swapping.