[EN]MODULE FOR AIRSENSOR 20-20-41 1 * 1 ST
Lieferant: SenseAnywhere
Module for AiroSensor 20-20-41, magnetic door contact set, 5 m cable length, magnet size 50L×14W×12D mm, 100 g weight (door contact set only; AiroSensor 20-20-41 not included)