Actinomycin D, Crystalline

Lieferant: Serva

10710.01 10710.02
SERA10710.01EA 75.6 EUR
SERA10710.01 SERA10710.02
Actinomycin D, Crystalline
Actinomycin D
Formel: C₆₂H₈₆N₁₂O₁₆
Molecular Weight: 1255.5 g/mol
MDL: MFCD00005033
CAS-Nummer: 50-76-0
UN: 3462
ADR: 6.1 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR