Adenosin
Lieferant: Serva
Synonyme:
9-β-D-Ribofuranosyladenine, (2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, Adenine-9-β-D-ribofuranoside, Adenine riboside
10770.02
SERA10770.02EA
0
EUR
SERA10770.02
Adenosin
Adenosin
Formel:
C₁₀H₁₃N₅O₄ Molecular Weight: 267,24 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00005752 CAS-Nummer: 58-61-7 EINECS: 200-389-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...