Tricain-Methansulfonat

Lieferant: Serva

Synonyme: TS 222, 3-Aminobenzoic acid Ethyl Ester Methanesulphonate, Tricaine, Ethyl 3-aminobenzoate methanesulphonate, Ethyl 3-aminobenzoate, methanesulphonic acid salt, Bio-Calm™, Ethyl-m-Aminobenzoate, Methanesulphonate salt, MS-222

12396.03
SERA12396.03EA 274 EUR
SERA12396.03
Tricain-Methansulfonat
Tricain-Methansulfonat
Formel: C₁₀H₁₅NO₅S
Molecular Weight: 261.29 g/mol
MDL: MFCD00013176
CAS-Nummer: 886-86-2
EINECS: 212-956-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR