Ampicillin

Lieferant: Serva

13397.01 13397.02 13397.03
SERA13397.01EA 63.1 EUR
SERA13397.01 SERA13397.02 SERA13397.03
Ampicillin
Ampicillin
Formel: C₁₆H₁₉N₃O₄S
Molecular Weight: 349.4 g/mol
MDL: MFCD00005175
CAS-Nummer: 69-53-4
EINECS: 200-709-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR