ABTS (Diammonium-2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazol-6-sulfonat])
Lieferant: Serva
|
Achtung
|
Synonyme:
ABTS, Ammonium 2,2'-(hydrazine-1,2-diylidene)bis(3-ethyl-2,3-dihydrobenzo[d]thiazole-6-sulphonate), 2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulphonic acid) diammonium salt
14364.01
SERA14364.01EA
126
EUR
SERA14364.01
ABTS (Diammonium-2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazol-6-sulfonat])
ABTS (Diammonium-2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazol-6-sulfonat])
Formel:
C₁₈H₂₄N₆O₆S₄ Molecular Weight: 548,68 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00010404 CAS-Nummer: 30931-67-0 EINECS: 250-396-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...