Benzethoniumchlorid
Lieferant: Serva
|
Gefahr
|
Synonyme:
Hyamine® 1622
14535.02
SERA14535.02EA
0
EUR
SERA14535.02
Benzethoniumchlorid
Benzethoniumchlorid
Formel:
C₂₇H₄₂ClNO₂ Molecular Weight: 448,09 g/mol Schmelzpunkt: 164…166 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011742 CAS-Nummer: 121-54-0 EINECS: 204-479-9 UN: 3077 ADR: 9,III Merck Index: 13,01074 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...