D-Cycloserin

Lieferant: Serva
Achtung

17685.01
SERA17685.01EA 0 EUR
SERA17685.01
D-Cycloserin
D-Cycloserin
Formel: C₃H₆N₂O₂
Lagertemperatur: Gefrierschrank
MDL: MFCD00005353
CAS-Nummer: 68-41-7
EINECS: 200-688-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR