Cytochalasin D
Lieferant: Serva
Synonyme:
7(S),18(R)-Dihydroxy-16(S),18-dimethyl-10-phenyl[11]cytochalasa-6(12),13(E),19(E)-trien-1,17-dion-21(R)-acetat
18017.01
SERA18017.01EA
0
EUR
SERA18017.01
Cytochalasin D
Cytochalasin D
Formel:
C₃₀H₃₇NO₆ Molecular Weight: 507,63 g/mol Schmelzpunkt: 259…260 °C |
MDL:
MFCD00077706 CAS-Nummer: 22144-77-0 EINECS: 244-804-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...