DEAE-Dextran

Lieferant: Serva

Synonyme: Diethylaminoethyldextran

18686.01 18686.02
SERA18686.01EA 0 EUR
SERA18686.01 SERA18686.02
DEAE-Dextran
DEAE-Dextran
Lagertemperatur: Raumtemperatur
MDL: MFCD00081561
CAS-Nummer: 9015-73-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR