Dextran, M.W. ~ 500,000

Lieferant: Serva

18695.03
SERA18695.03EA 0 EUR
SERA18695.03
Dextran, M.W. ~ 500,000
Dextran
Formel: (C₆H₁₀O₅)n
MDL: MFCD00130935
CAS-Nummer: 9004-54-0
EINECS: 232-677-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR