n-Dodecyl-β-D-maltosid

Lieferant: Serva
Achtung

Synonyme: DDM

20780.01 20780.02
SERA20780.01EA 0 EUR
SERA20780.01 SERA20780.02
n-Dodecyl-β-D-maltosid
n-Dodecyl-β-D-maltosid
Formel: C₂₄H₄₆O₁₁
Molecular Weight: 510.6 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00043012
CAS-Nummer: 69227-93-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR