Fluoresceindiacetat

Lieferant: Serva

21575.02
SERA21575.02EA 0 EUR
SERA21575.02
Fluoresceindiacetat
Fluoresceindiacetat
Formel: C₂₄H₁₆O₇
MDL: MFCD00005062
CAS-Nummer: 596-09-8
EINECS: 209-877-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR